3,5,6-Trichlorpicolinsäure

Lieferant: ACTIVATE SCIENTIFIC

AS36031G1 AS36031G5
ACTIAS36031G1EA 0 CHF
ACTIAS36031G1 ACTIAS36031G5
3,5,6-Trichlorpicolinsäure
3,5,6-Trichlorpicolinsäure
Formel: C₆H₂Cl₃NO₂
MW: 226.4446 g/mol
MDL Number: MFCD01086756
CAS-Nummer: 40360-44-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR