3,4-Dichlorpicolinsäure

Lieferant: ACTIVATE SCIENTIFIC

AS23673G1 AS23673G5
ACTIAS23673G1EA 0 CHF
ACTIAS23673G1 ACTIAS23673G5
3,4-Dichlorpicolinsäure
3,4-Dichlorpicolinsäure
Formel: C₆H₃Cl₂NO₂
MDL Number: MFCD01861980
CAS-Nummer: 959578-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR