4,6-Dichlorisatin

Lieferant: ACTIVATE SCIENTIFIC

AS30500G1 AS30500G5
ACTIAS30500G1EA 0 CHF
ACTIAS30500G1 ACTIAS30500G5
4,6-Dichlorisatin
4,6-Dichlorisatin
Formel: C₈H₃Cl₂NO₂
MDL Number: MFCD01971146
CAS-Nummer: 18711-15-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR