4-Chlor-2-pyridincarbonitril

Lieferant: ACTIVATE SCIENTIFIC

AS8328G1 AS8328G5
ACTIAS8328G1EA 0 CHF
ACTIAS8328G1 ACTIAS8328G5
4-Chlor-2-pyridincarbonitril
4-Chlor-2-pyridincarbonitril
Formel: C₆H₃ClN₂
MDL Number: MFCD06009833
CAS-Nummer: 19235-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR