4-Brom-3,5-xylidin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Bromo-3,5-dimethylaniline

AS35377G1 AS35377G5
ACTIAS35377G1EA 0 CHF
ACTIAS35377G1 ACTIAS35377G5
4-Brom-3,5-xylidin
4-Brom-3,5-xylidin
Formel: C₈H₁₀BrN
MW: 200.0800 g/mol
MDL Number: MFCD07780648
CAS-Nummer: 59557-90-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR