15-Hydroxypentadecansäure

Lieferant: MATREYA

MAYA1881EA 0 CHF
MAYA1881
15-Hydroxypentadecansäure
15-Hydroxypentadecanoic acid
Formel: C₁₅H₃₀O₃
MW: 258,40 g/mol
MDL Number: MFCD00039535
CAS-Nummer: 4617-33-8
EINECS: 225-026-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR