10-Bromdecansäure

Lieferant: ACTIVATE SCIENTIFIC

AS25320G100 AS25320G25
ACTIAS25320G100EA 0 CHF
ACTIAS25320G100 ACTIAS25320G25
10-Bromdecansäure
10-Bromodecanoic acid
Formel: C₁₀H₁₉BrO₂
MDL Number: MFCD00014388
CAS-Nummer: 50530-12-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR