4-(1-Pyrrolidinylcarbonyl)phenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-(1-Pyrrolidinylcarbonyl)benzeneboronic acid, 4-(Pyrrolidine-1-carbonyl)phenylboronic acid, (4-(Pyrrolidine-1-carbonyl)phenyl)boronic acid

AS50996G1 AS50996G5
ACTIAS50996G1EA 0 CHF
ACTIAS50996G1 ACTIAS50996G5
4-(1-Pyrrolidinylcarbonyl)phenylboronsäure
4-(1-Pyrrolidinylcarbonyl)phenylboronsäure
Formel: C₁₁H₁₄BNO₃
MDL Number: MFCD03411950
CAS-Nummer: 389621-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR