3-(1-Pyrrolidinylcarbonyl)phenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (3-(Pyrrolidine-1-carbonyl)phenyl)boronic acid

AS51548G1 AS51548G5
ACTIAS51548G1EA 0 CHF
ACTIAS51548G1 ACTIAS51548G5
3-(1-Pyrrolidinylcarbonyl)phenylboronsäure
3-(1-Pyrrolidinylcarbonyl)phenylboronsäure
Formel: C₁₁H₁₄BNO₃
MW: 219.0500 g/mol
MDL Number: MFCD04115685
CAS-Nummer: 723281-53-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR