4-Brom-1-(2-tetrahydropyranyl)-1H-pyrazol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazole, 4-Bromo-1-(tetrahydro-2H-pyran-2-yl-1H)-pyrazole

AS35084G1 AS35084G10 AS35084G5
ACTIAS35084G1EA 0 CHF
ACTIAS35084G1 ACTIAS35084G10 ACTIAS35084G5
4-Brom-1-(2-tetrahydropyranyl)-1H-pyrazol
4-Brom-1-(2-tetrahydropyranyl)-1H-pyrazol
Formel: C₈H₁₁BrN₂O
MW: 231,0900 g/mol
MDL Number: MFCD12026052
CAS-Nummer: 82099-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR