1-(4-Chlorphenyl)cyclobutancarbonsäure

Lieferant: ACTIVATE SCIENTIFIC

AS34923G1 AS34923G5
ACTIAS34923G1EA 0 CHF
ACTIAS34923G1 ACTIAS34923G5
1-(4-Chlorphenyl)cyclobutancarbonsäure
1-(4-Chlorophenyl)cyclobutanecarboxylic acid
Formel: C₁₁H₁₁ClO₂
MDL Number: MFCD00001324
CAS-Nummer: 50921-39-6
EINECS: 256-847-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR