3-Chlor-4-morpholinobenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS21510G1 AS21510G25 AS21510G5
ACTIAS21510G1EA 148 CHF
ACTIAS21510G1 ACTIAS21510G25 ACTIAS21510G5
3-Chlor-4-morpholinobenzaldehyd
3-Chlor-4-morpholinobenzaldehyd
Formel: C₁₁H₁₂ClNO₂
MW: 225,6700 g/mol
MDL Number: MFCD06740107
CAS-Nummer: 886500-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR