2-Morpholinobenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS20321G1 AS20321G100 AS20321G25 AS20321G5
ACTIAS20321G1EA 0 CHF
ACTIAS20321G1 ACTIAS20321G100 ACTIAS20321G25 ACTIAS20321G5
2-Morpholinobenzaldehyd
2-Morpholinobenzaldehyd
Formel: C₁₁H₁₃NO₂
MW: 191,2300 g/mol
MDL Number: MFCD00662562
CAS-Nummer: 58028-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR