4-Brom-2-morpholinobenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS33012G1 AS33012G5
ACTIAS33012G1EA 0 CHF
ACTIAS33012G1 ACTIAS33012G5
4-Brom-2-morpholinobenzaldehyd
4-Brom-2-morpholinobenzaldehyd
Formel: C₁₁H₁₂BrNO₂
MDL Number: MFCD06656558
CAS-Nummer: 736990-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR