7-Chlor-4-chinazolon

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 7-Chloro-4-hydroxyquinazoline

AS20008G1 AS20008G25 AS20008G5
ACTIAS20008G1EA 23 CHF
ACTIAS20008G1 ACTIAS20008G25 ACTIAS20008G5
7-Chlor-4-chinazolon
7-Chlor-4-chinazolon
Formel: C₈H₅ClN₂O
MW: 180,5900 g/mol
MDL Number: MFCD01570890
CAS-Nummer: 31374-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR