7-Bromchinazolin

Lieferant: ACTIVATE SCIENTIFIC

AS2469G1 AS2469G5
ACTIAS2469G1EA 97 CHF
ACTIAS2469G1 ACTIAS2469G5
7-Bromchinazolin
7-Bromoquinazoline
Formel: C₈H₅BrN₂
MW: 209.0400 g/mol
MDL Number: MFCD07778500
CAS-Nummer: 89892-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR