6-Bromchinazolin

Lieferant: ACTIVATE SCIENTIFIC

AS2465G1 AS2465G5
ACTIAS2465G1EA 34 CHF
ACTIAS2465G1 ACTIAS2465G5
6-Bromchinazolin
6-Bromoquinazoline
Formel: C₈H₅BrN₂
MW: 209,0400 g/mol
MDL Number: MFCD08236695
CAS-Nummer: 89892-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR