5-Cyan-2-thiophenboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (5-Cyanothiophen-2-yl)boronic acid, 5-Cyanothiophene-2-boronic acid

AS50167G1 AS50167G5
ACTIAS50167G1EA 0 CHF
ACTIAS50167G1 ACTIAS50167G5
5-Cyan-2-thiophenboronsäure
5-Cyano-2-thiopheneboronic acid
Formel: C₅H₄BNO₂S
MDL Number: MFCD02094029
CAS-Nummer: 305832-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR