4-Brom-2,6-dimethylpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS33830G1 AS33830G5
ACTIAS33830G1EA 39.57 CHF
ACTIAS33830G1 ACTIAS33830G5
4-Brom-2,6-dimethylpyridin
4-Bromo-2,6-dimethylpyridine
Formel: C₇H₈BrN
MW: 186.0491 g/mol
MDL Number: MFCD00671475
CAS-Nummer: 5093-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR