2-Brom-4,6-dimethylpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS33433G1 AS33433G5
ACTIAS33433G1EA 0 CHF
ACTIAS33433G1 ACTIAS33433G5
2-Brom-4,6-dimethylpyridin
2-Bromo-4,6-dimethylpyridine
Formel: C₇H₈BrN
MW: 186.05 g/mol
MDL Number: MFCD00082591
CAS-Nummer: 4926-26-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR