3-Nitro-4-picolin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Methyl-3-nitropyridine

AS31663G1 AS31663G5
ACTIAS31663G1EA 0 CHF
ACTIAS31663G1 ACTIAS31663G5
3-Nitro-4-picolin
3-Nitro-4-picolin
Formel: C₆H₆N₂O₂
MW: 138,13 g/mol
MDL Number: MFCD00051829
CAS-Nummer: 5832-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR