2-Nitro-4-picolin

Lieferant: ACTIVATE SCIENTIFIC

AS1534G1 AS1534G5
ACTIAS1534G1EA 0 CHF
ACTIAS1534G1 ACTIAS1534G5
2-Nitro-4-picolin
2-Nitro-4-picolin
Formel: C₆H₆N₂O₂
MDL Number: MFCD04114155
CAS-Nummer: 18368-71-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR