3-Amino-2-methoxy-6-picolin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Amino-2-methoxy-6-methylpyridine

AS31785G1 AS31785G5
ACTIAS31785G1EA 0 CHF
ACTIAS31785G1 ACTIAS31785G5
3-Amino-2-methoxy-6-picolin
3-Amino-2-methoxy-6-picolin
Formel: C₇H₁₀N₂O
MDL Number: MFCD03095079
CAS-Nummer: 186413-79-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR