3-Amino-2-methoxy-4-picolin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Methoxy-4-methylpyridin-3-amine

AS32603G1 AS32603G5
ACTIAS32603G1EA 0 CHF
ACTIAS32603G1 ACTIAS32603G5
3-Amino-2-methoxy-4-picolin
3-Amino-2-methoxy-4-picolin
Formel: C₇H₁₀N₂O
MW: 138,1671 g/mol
MDL Number: MFCD08277287
CAS-Nummer: 76005-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR