5-Acetyl-2-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS33428G1 AS33428G5
ACTIAS33428G1EA 0 CHF
ACTIAS33428G1 ACTIAS33428G5
5-Acetyl-2-methylpyridin
5-Acetyl-2-methylpyridine
Formel: C₈H₉NO
MW: 135,17 g/mol
MDL Number: MFCD00128113
CAS-Nummer: 36357-38-7
EINECS: 252-995-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR