3-Amino-2-cyan-5-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS35373G1 AS35373G10 AS35373G5
ACTIAS35373G1EA 0 CHF
ACTIAS35373G1 ACTIAS35373G10 ACTIAS35373G5
3-Amino-2-cyan-5-methylpyridin
3-Amino-2-cyano-5-methylpyridine
Formel: C₇H₇N₃
MDL Number: MFCD11227185
CAS-Nummer: 1001635-30-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR