6-Amino-3-cyan-2-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 6-Amino-2-methylnicotinonitrile, 6-Amino-3-cyano-2-methylpyridine, 6-Amino-2-methylpyridine-3-carbonitrile

AS50797G1 AS50797G5
ACTIAS50797G1EA 0 CHF
ACTIAS50797G1 ACTIAS50797G5
6-Amino-3-cyan-2-methylpyridin
6-Amino-3-cyan-2-methylpyridin
Formel: C₇H₇N₃
MDL Number: MFCD05865116
CAS-Nummer: 183428-90-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR