2-Amino-5-cyan-3-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 6-Amino-5-methylnicotinonitrile, 2-Amino-3-methyl-5-cyanopyridine

AS1744G1 AS1744G5
ACTIAS1744G1EA 0 CHF
ACTIAS1744G1 ACTIAS1744G5
2-Amino-5-cyan-3-methylpyridin
2-Amino-5-cyano-3-methylpyridine
Formel: C₇H₇N₃
MW: 133.1506 g/mol
MDL Number: MFCD06637357
CAS-Nummer: 183428-91-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR