3-Chlor-5-(trifluormethyl)picolinonitril

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Chloro-5-(trifluoromethyl)pyridine-2-carbonitrile

AS34847G1 AS34847G5
ACTIAS34847G1EA 0 CHF
ACTIAS34847G1 ACTIAS34847G5
3-Chlor-5-(trifluormethyl)picolinonitril
3-Chlor-5-(trifluormethyl)picolinonitril
Formel: C₇H₂ClF₃N₂
MW: 206.5524 g/mol
MDL Number: MFCD00664066
CAS-Nummer: 80194-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR