2-Chlor-3-cyan-6-picolin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Chloro-3-cyano-6-methylpyridine, 2-Chloro-6-methylnicotinonitrile

AS33661G1 AS33661G10 AS33661G5
ACTIAS33661G1EA 0 CHF
ACTIAS33661G1 ACTIAS33661G10 ACTIAS33661G5
2-Chlor-3-cyan-6-picolin
2-Chlor-3-cyan-6-picolin
Formel: C₇H₅ClN₂
MW: 152,58 g/mol
MDL Number: MFCD00082767
CAS-Nummer: 28900-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR