3-Brom-5-methylpicolinsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Bromo-5-methylpyridine-2-carboxylic acid

AS36078G1 AS36078G5
ACTIAS36078G1EA 0 CHF
ACTIAS36078G1 ACTIAS36078G5
3-Brom-5-methylpicolinsäure
3-Brom-5-methylpicolinsäure
Formel: C₇H₆BrNO₂
MW: 216.0320 g/mol
MDL Number: MFCD17214206
CAS-Nummer: 1211515-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR