2-Acetyl-5-brompyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1-(5-Bromopyridin-2-yl)ethanone

AS31710G1 AS31710G5
ACTIAS31710G1EA 0 CHF
ACTIAS31710G1 ACTIAS31710G5
2-Acetyl-5-brompyridin
2-Acetyl-5-brompyridin
Formel: C₇H₆BrNO
MW: 200,0300 g/mol
MDL Number: MFCD04974523
CAS-Nummer: 214701-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR