3-Acetyl-5-brompyridin

Lieferant: ACTIVATE SCIENTIFIC

AS31273G1 AS31273G5
ACTIAS31273G1EA 0 CHF
ACTIAS31273G1 ACTIAS31273G5
3-Acetyl-5-brompyridin
3-Acetyl-5-brompyridin
Formel: C₇H₆BrNO
MDL Number: MFCD03086033
CAS-Nummer: 38940-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR