5-Acetyl-2-brompyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Acetyl-6-bromopyridine

AS50474G1 AS50474G5
ACTIAS50474G1EA 0 CHF
ACTIAS50474G1 ACTIAS50474G5
5-Acetyl-2-brompyridin
5-Acetyl-2-brompyridin
Formel: C₇H₆BrNO
MDL Number: MFCD04974527
CAS-Nummer: 139042-59-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR