3-Acetyl-2-brompyridin

Lieferant: ACTIVATE SCIENTIFIC

AS31865G1 AS31865G5
ACTIAS31865G1EA 0 CHF
ACTIAS31865G1 ACTIAS31865G5
3-Acetyl-2-brompyridin
3-Acetyl-2-brompyridin
Formel: C₇H₆BrNO
MW: 200.0300 g/mol
MDL Number: MFCD04114370
CAS-Nummer: 84199-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR