5-Brom-2-cyan-3-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS35744G1 AS35744G5
ACTIAS35744G1EA 0 CHF
ACTIAS35744G1 ACTIAS35744G5
5-Brom-2-cyan-3-methylpyridin
5-Brom-2-cyan-3-methylpyridin
Formel: C₇H₅BrN₂
MDL Number: MFCD11975788
CAS-Nummer: 156072-86-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR