3-Brom-2-cyan-5-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Bromo-5-methylpicolinonitrile, 3-Bromo-5-methylpyridine-2-carbonitrile

AS36317G1 AS36317G5
ACTIAS36317G1EA 0 CHF
ACTIAS36317G1 ACTIAS36317G5
3-Brom-2-cyan-5-methylpyridin
3-Bromo-2-cyano-5-methylpyridine
Formel: C₇H₅BrN₂
MDL Number: MFCD16988815
CAS-Nummer: 474824-78-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR