2-Fluor-4-iodnicotinsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Fluoro-4-iodo-nicotinic acid

AS31991G1 AS31991G5
ACTIAS31991G1EA 0 CHF
ACTIAS31991G1 ACTIAS31991G5
2-Fluor-4-iodnicotinsäure
2-Fluor-4-iodnicotinsäure
Formel: C₆H₃FINO₂
MW: 267,0000 g/mol
MDL Number: MFCD03095310
CAS-Nummer: 884494-51-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR