5-Brom-2-(methylthio)pyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Bromo-2-methylthiopyridine, 2-Methylthio-5-bromopyridine

AS51250G1 AS51250G5
ACTIAS51250G1EA 0 CHF
ACTIAS51250G1 ACTIAS51250G5
5-Brom-2-(methylthio)pyridin
5-Brom-2-(methylthio)pyridin
Formel: C₆H₆BrNS
MW: 204,0875 g/mol
MDL Number: MFCD03411571
CAS-Nummer: 51933-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR