2-(Tetrahydro-2H-pyran-4-yloxy)pyridin-5-boronsäurepinakolester

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-(Tetrahydropyran-4-yloxy)-5-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)pyridine

AS32641G1 AS32641G5
ACTIAS32641G1EA 0 CHF
ACTIAS32641G1 ACTIAS32641G5
2-(Tetrahydro-2H-pyran-4-yloxy)pyridin-5-boronsäurepinakolester
2-(Tetrahydro-2H-pyran-4-yloxy)pyridin-5-boronsäurepinakolester
Formel: C₁₆H₂₄BNO₄
MW: 305,18 g/mol
MDL Number: MFCD09702390
CAS-Nummer: 910036-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR