Tetrahydro-2H-pyran-4-carbonylchlorid

Lieferant: ACTIVATE SCIENTIFIC

AS1353G1 AS1353G5
ACTIAS1353G1EA 0 CHF
ACTIAS1353G1 ACTIAS1353G5
Tetrahydro-2H-pyran-4-carbonylchlorid
Tetrahydro-2H-pyran-4-carbonylchlorid
Formel: C₆H₉ClO₂
MW: 148,5875 g/mol
MDL Number: MFCD06200863
CAS-Nummer: 40191-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR