8-Fluorchinolin

Lieferant: ACTIVATE SCIENTIFIC

AS21399G1 AS21399G25 AS21399G5
ACTIAS21399G1EA 23 CHF
ACTIAS21399G1 ACTIAS21399G25 ACTIAS21399G5
8-Fluorchinolin
8-Fluoroquinoline
Formel: C₉H₆FN
MW: 147,1500 g/mol
MDL Number: MFCD01685514
CAS-Nummer: 394-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR