6-Fluorchinolin

Lieferant: ACTIVATE SCIENTIFIC

AS21400G1 AS21400G25 AS21400G5
ACTIAS21400G1EA 0 CHF
ACTIAS21400G1 ACTIAS21400G25 ACTIAS21400G5
6-Fluorchinolin
6-Fluoroquinoline
Formel: C₉H₆FN
MW: 147.15 g/mol
MDL Number: MFCD01685512
CAS-Nummer: 396-30-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR