5,7-Difluorchinolin

Lieferant: ACTIVATE SCIENTIFIC

AS35662G1 AS35662G5
ACTIAS35662G1EA 0 CHF
ACTIAS35662G1 ACTIAS35662G5
5,7-Difluorchinolin
5,7-Difluoroquinoline
Formel: C₉H₅F₂N
MW: 165,1400 g/mol
MDL Number: MFCD00474638
CAS-Nummer: 34522-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR