4'-Chlorbiphenyl-4-ylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4'-Chloro-4-biphenylboronic acid, (4'-Chloro-[1,1'-biphenyl]-4-yl)boronic acid

AS36417G1 AS36417G5
ACTIAS36417G1EA 0 CHF
ACTIAS36417G1 ACTIAS36417G5
4'-Chlorbiphenyl-4-ylboronsäure
4'-Chlorbiphenyl-4-ylboronsäure
Formel: C₁₂H₁₀BClO₂
MW: 232,4700 g/mol
MDL Number: MFCD08544389
CAS-Nummer: 364044-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR