(±)-Cbz-α-phosphonoglycintrimethylester

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: N-Benzyloxycarbonyl-2-phosphonoglycine trimethyl ester, (±)-Benzyloxycarbonyl-α-phosphonoglycine trimethyl ester, (±)-Methyl 2-benzyloxycarbonylamino-2-(dimethoxyphosphinyl)acetate, Methyl 2-{[(benzyloxy)carbonyl]amino}-2-(dimethoxyphosphoryl)acetate, Methyl 2-(((benzyloxy)carbonyl)amino)-2-(dimethoxyphosphoryl)acetate

AS12627G100 AS12627G25
ACTIAS12627G100EA 0 CHF
ACTIAS12627G100 ACTIAS12627G25
(±)-Cbz-α-phosphonoglycintrimethylester
(±)-Cbz-α-phosphonoglycintrimethylester
Formel: C₁₃H₁₈NO₇P
MDL Number: MFCD00043304
CAS-Nummer: 88568-95-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR