3-Amino-5-nitrobenzylalkohol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (3-Amino-5-nitrophenyl)methanol

AS32513G1 AS32513G5
ACTIAS32513G1EA 0 CHF
ACTIAS32513G1 ACTIAS32513G5
3-Amino-5-nitrobenzylalkohol
3-Amino-5-nitrobenzylalkohol
Formel: C₇H₈N₂O₃
MDL Number: MFCD08236778
CAS-Nummer: 90390-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR