2-Amino-4-methylbenzamid

Lieferant: ACTIVATE SCIENTIFIC

AS35485G1 AS35485G5
ACTIAS35485G1EA 0 CHF
ACTIAS35485G1 ACTIAS35485G5
2-Amino-4-methylbenzamid
2-Amino-4-methylbenzamid
Formel: C₈H₁₀N₂O
MW: 150,1800 g/mol
MDL Number: MFCD00221474
CAS-Nummer: 39549-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR