N-Boc-DL-phenylalaninol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Boc-DL-Phenylalaninol, tert-Butyl (1-hydroxy-3-phenylpropan-2-yl)carbamate

AS22376G25 AS22376G5 AS22376G1
ACTIAS22376G25EA 0 CHF
ACTIAS22376G25 ACTIAS22376G5 ACTIAS22376G1
N-Boc-DL-phenylalaninol
N-Boc-DL-phenylalaninol
Formel: C₁₄H₂₁NO₃
MDL Number: MFCD01457313
CAS-Nummer: 145149-48-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR