2-Amino-3-chlorbenzylalkohol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (2-Amino-3-chlorophenyl)methanol

AS32038G1 AS32038G5
ACTIAS32038G1EA 0 CHF
ACTIAS32038G1 ACTIAS32038G5
2-Amino-3-chlorbenzylalkohol
2-Amino-3-chlorbenzylalkohol
Formel: C₇H₈ClNO
MDL Number: MFCD13193432
CAS-Nummer: 61487-25-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR